missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylphenol, 99+%, Thermo Scientific Chemicals

Product Code. 10162420
Click to view available options
Quantity:
5 g
100 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-50-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00002323
InChI Key CFKMVGJGLGKFKI-UHFFFAOYSA-N
Synonym chlorocresol, 4-chloro-m-cresol, baktol, parol, p-chloro-m-cresol, ottafact, phenol, 4-chloro-3-methyl, 4-chloro-3-cresol, candaseptic, baktolan
PubChem CID 1732
ChEBI CHEBI:34395
IUPAC Name 4-chloro-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 63.0°C to 65.0°C
Color White
Boiling Point 242°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula ClC6H3(CH3)OH
Quantity 500 g
Beilstein 06, 381
Merck Index 15, 2133
Solubility Information Solubility in water: 4g/L (20 c). Other solubilities: soluble in alkalies,soluble in most common organic solvents
Formula Weight 142.58
Percent Purity 99+%
Physical Form Tablets
Chemical Name or Material 4-Chloro-3-methylphenol, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-3-methylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.