missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrophenol, 99%, Thermo Scientific Chemicals

42.70 € - 140.00 €

Chemical Identifiers

CAS 610-78-6
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.552
MDL Number MFCD00043546
InChI Key JUIKCULGDIZNDI-UHFFFAOYSA-N
PubChem CID 69127
IUPAC Name 4-chloro-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388225
View Documents
Thermo Scientific Alfa Aesar
L10108.06
5 g
42.70 €
5g
Please to purchase this item. Need a web account? Register with us today!
11398225
View Documents
Thermo Scientific Alfa Aesar
L10108.14
25 g
140.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

610-78-6
173.552
JUIKCULGDIZNDI-UHFFFAOYSA-N
4-chloro-3-nitrophenol
C6H4ClNO3
MFCD00043546
69127
C1=CC(=C(C=C1O)[N+](=O)[O-])Cl

Specifications

610-78-6
Characteristic
MFCD00043546
phenol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenol, 4chloro-3-nitrophenol, pubchem10459, 4-chlor-3-nitro-phenol, 3-nitro-4-chloro phenol, acmc-1azj5, phenol,4-chloro-3-nitro, 4-chloranyl-3-nitro-phenol, 2-chloro-5-hydroxy nitrobenzene
C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
173.552
173.56
4-Chloro-3-nitrophenol
124°C to 129°C
C6H4ClNO3
2094546
JUIKCULGDIZNDI-UHFFFAOYSA-N
4-chloro-3-nitrophenol
69127
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only