missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-3-nitrophenyl isothiocyanate, Thermo Scientific™

Product Code. 10506035
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 127142-66-9
Molecular Formula C7H3ClN2O2S
Molecular Weight (g/mol) 214.623
InChI Key ZXGZBHIDSJXKLE-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrophenyl isothiocyanate, 4-chloro-3-nitrophenylisothiocyanate, acmc-1c0qb, 4-chloro-3-nitrobenzenisothiocyanate, #, benzene,1-chloro-4-isothiocyanato-2-nitro, 1-chloranyl-4-isothiocyanato-2-nitro-benzene
PubChem CID 145581
IUPAC Name 1-chloro-4-isothiocyanato-2-nitrobenzene
SMILES C1=CC(=C(C=C1N=C=S)[N+](=O)[O-])Cl

Specifications

Assay Percent Range 97%
Quantity 5 g
Formula Weight 214.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-nitrophenyl isothiocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.