missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-4'-fluorobutyrophenone, 96%, Thermo Scientific Chemicals

28.34 € - 63.90 €

Chemical Identifiers

CAS 3874-54-2
Molecular Formula C10H10ClFO
Molecular Weight (g/mol) 200.637
MDL Number MFCD00001007
InChI Key HXAOUYGZEOZTJO-UHFFFAOYSA-N
Synonym 4-chloro-4'-fluorobutyrophenone, 4-chloro-1-4-fluorophenyl butan-1-one, 3-4-fluorobenzoyl propyl chloride, 4-fluorobenzoylpropyl chloride, 4-chloro-p-fluorobutyrophenone, 3-chloropropyl 4-fluorophenyl ketone, 1-butanone, 4-chloro-1-4-fluorophenyl, 4'-fluoro-4-chlorobutyrophenone, p-fluorobenzoylpropyl chloride, p-fluoro-4-chlorobutyrophenone
PubChem CID 19750
IUPAC Name 4-chloro-1-(4-fluorophenyl)butan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11429013
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11121.14
25 g
28.34 €
25g
Please to purchase this item. Need a web account? Register with us today!
11439013
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A11121.22
100 g
63.90 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3874-54-2
200.637
HXAOUYGZEOZTJO-UHFFFAOYSA-N
19750
C1=CC(=CC=C1C(=O)CCCCl)F
C10H10ClFO
MFCD00001007
4-chloro-4'-fluorobutyrophenone, 4-chloro-1-4-fluorophenyl butan-1-one, 3-4-fluorobenzoyl propyl chloride, 4-fluorobenzoylpropyl chloride, 4-chloro-p-fluorobutyrophenone, 3-chloropropyl 4-fluorophenyl ketone, 1-butanone, 4-chloro-1-4-fluorophenyl, 4'-fluoro-4-chlorobutyrophenone, p-fluorobenzoylpropyl chloride, p-fluoro-4-chlorobutyrophenone
4-chloro-1-(4-fluorophenyl)butan-1-one

Specifications

3874-54-2
1.23
152°C (306°F)
1.5255
608741
HXAOUYGZEOZTJO-UHFFFAOYSA-N
4-chloro-1-(4-fluorophenyl)butan-1-one
19750
96%
5°C to 6°C
120°C to 122°C (0.7 mmHg)
C10H10ClFO
MFCD00001007
4-chloro-4'-fluorobutyrophenone, 4-chloro-1-4-fluorophenyl butan-1-one, 3-4-fluorobenzoyl propyl chloride, 4-fluorobenzoylpropyl chloride, 4-chloro-p-fluorobutyrophenone, 3-chloropropyl 4-fluorophenyl ketone, 1-butanone, 4-chloro-1-4-fluorophenyl, 4'-fluoro-4-chlorobutyrophenone, p-fluorobenzoylpropyl chloride, p-fluoro-4-chlorobutyrophenone
C1=CC(=CC=C1C(=O)CCCCl)F
200.637
200.64
4-Chloro-4'-fluorobutyrophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319

missing translation for 'einecsNumber' : 223-395-3

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

Product Identifier
  • 4-Chloro-4'-fluorobutyrophenone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : 4-Chloro-4'-fluorobutyrophenone

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only