missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-4'-fluorobutyrophenone, 97%, Thermo Scientific Chemicals

25.40 € - 210.05 €

Chemical Identifiers

CAS 3874-54-2
Molecular Formula C10H10ClFO
Molecular Weight (g/mol) 200.64
MDL Number MFCD00001007
InChI Key HXAOUYGZEOZTJO-UHFFFAOYSA-N
Synonym 4-chloro-4'-fluorobutyrophenone, 4-chloro-1-4-fluorophenyl butan-1-one, 3-4-fluorobenzoyl propyl chloride, 4-fluorobenzoylpropyl chloride, 4-chloro-p-fluorobutyrophenone, 3-chloropropyl 4-fluorophenyl ketone, 1-butanone, 4-chloro-1-4-fluorophenyl, 4'-fluoro-4-chlorobutyrophenone, p-fluorobenzoylpropyl chloride, p-fluoro-4-chlorobutyrophenone
PubChem CID 19750
IUPAC Name 4-chloro-1-(4-fluorophenyl)butan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)F
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10375690
View Documents
Thermo Scientific Acros
109230250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10733831
View Documents
Thermo Scientific Acros
109231000
100 g Glass bottle
67.08 €
100g
Please to purchase this item. Need a web account? Register with us today!
10122710
View Documents
Thermo Scientific Acros
109235000
500 g Glass bottle
210.05 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3874-54-2
200.64
HXAOUYGZEOZTJO-UHFFFAOYSA-N
19750
C1=CC(=CC=C1C(=O)CCCCl)F
C10H10ClFO
MFCD00001007
4-chloro-4'-fluorobutyrophenone, 4-chloro-1-4-fluorophenyl butan-1-one, 3-4-fluorobenzoyl propyl chloride, 4-fluorobenzoylpropyl chloride, 4-chloro-p-fluorobutyrophenone, 3-chloropropyl 4-fluorophenyl ketone, 1-butanone, 4-chloro-1-4-fluorophenyl, 4'-fluoro-4-chlorobutyrophenone, p-fluorobenzoylpropyl chloride, p-fluoro-4-chlorobutyrophenone
4-chloro-1-(4-fluorophenyl)butan-1-one

Specifications

3874-54-2
100.0
Green-Yellow
130.0°C to 132.0°C (1.3 mbar)
Authentic
C10H10ClFO
1.5245 to 1.5265
07,IV,711
4-chloro-4'-fluorobutyrophenone, 4-chloro-1-4-fluorophenyl butan-1-one, 3-4-fluorobenzoyl propyl chloride, 4-fluorobenzoylpropyl chloride, 4-chloro-p-fluorobutyrophenone, 3-chloropropyl 4-fluorophenyl ketone, 1-butanone, 4-chloro-1-4-fluorophenyl, 4'-fluoro-4-chlorobutyrophenone, p-fluorobenzoylpropyl chloride, p-fluoro-4-chlorobutyrophenone
HXAOUYGZEOZTJO-UHFFFAOYSA-N
4-chloro-1-(4-fluorophenyl)butan-1-one
19750
97%
4-Chloro-4'-fluorobutyrophenone, 97%
96.0
5.0°C to 6.0°C
1.2300g/mL
152°C
96% min. (GC)
FC6H4CO(CH2)3Cl
MFCD00001007
1.23
Solubility in water: 0.38g/L water (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,n,n-dmf,,acetone and dichloromethane
C1=CC(=CC=C1C(=O)CCCCl)F
200.64
200.64
Oil
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 223-395-3

missing translation for 'tsca' : TSCA

Recommended Storage : The substance may darken on ageing

Product Identifier
  • 4-Chloro-4'-fluorobutyrophenone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only