missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-alpha-methylphenylacetic acid, 97%, Thermo Scientific Chemicals

Product Code. 11312336
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-alpha-methylphenylacetic acid is used to produce a-(4-Chlor-3-nitrophenyl)propionsaure. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-95-4
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00044670
InChI Key YOZILQVNIWNPFP-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl propanoic acid, 4-chloro-alpha-methylphenylacetic acid, dl-2-4-chlorophenyl propan-oic acid, dl-p-chloro-.alpha.-methylphenylacetic acid, benzeneacetic acid, 4-chloro-.alpha.-methyl, dl-2-4-chlorophenyl propanoic acid, 2-4-chloro-phenyl-propionic acid, dl-4-chloro-alpha-methylphenylacetic acid, ?-p-chlorohydratropic acid, 4-chloro methylphenylacetic acid
PubChem CID 102525
IUPAC Name 2-(4-chlorophenyl)propanoic acid
SMILES CC(C1=CC=C(C=C1)Cl)C(=O)O

Specifications

Melting Point 55°C to 59°C
Boiling Point 121°C to 126°C (0.2 mmHg)
Quantity 5 g
Beilstein 2328710
Solubility Information Soluble in water.
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material 4-Chloro-alpha-methylphenylacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.