missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide, 97%, Thermo Scientific™

50.90 € - 599.00 €

Chemical Identifiers

CAS 41859-57-8
Molecular Formula C15H14ClNO2
Molecular Weight (g/mol) 275.73
MDL Number MFCD00218513
InChI Key ZTLWJYCDAXUIBK-UHFFFAOYSA-N
Synonym n-4-chlorobenzoyl-tyramine, 4-chloro-n-4-hydroxyphenethyl benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl benzamide, n-4-chlorobenzoyl tyramine, benzamide, p-chloro-n-2-p-hydroxyphenyl ethyl, p-chloro-n-2-p-hydroxyphenyl ethyl benzamide, benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl, bez-daShow MoreShow Less
PubChem CID 198234
IUPAC Name 4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
SMILES OC1=CC=C(CCNC(=O)C2=CC=C(Cl)C=C2)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15411076
SDS View Documents
Thermo Scientific Alfa Aesar
H54344.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15491066
SDS View Documents
Thermo Scientific Alfa Aesar
H54344.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15401076
SDS View Documents
Thermo Scientific Alfa Aesar
H54344.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 255-566-3

missing translation for 'rtecsNumber' : CV2450481

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.