missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

66.63 € - 948.72 €

Chemical Identifiers

CAS 95-83-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00011691
InChI Key BXIXXXYDDJVHDL-UHFFFAOYSA-N
Synonym 4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
PubChem CID 7263
ChEBI CHEBI:82301
IUPAC Name 4-chlorobenzene-1,2-diamine
SMILES NC1=CC=C(Cl)C=C1N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11449772
View Documents
Thermo Scientific Alfa Aesar
A10823.14
25 g
66.63 €
25g
Please to purchase this item. Need a web account? Register with us today!
11459772
View Documents
Thermo Scientific Alfa Aesar
A10823.22
100 g
188.94 €
100g
Please to purchase this item. Need a web account? Register with us today!
11469772
View Documents
Thermo Scientific Alfa Aesar
A10823.36
500 g
948.72 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95-83-0
142.59
BXIXXXYDDJVHDL-UHFFFAOYSA-N
7263
4-chlorobenzene-1,2-diamine
C6H7ClN2
MFCD00011691
4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
CHEBI:82301
NC1=CC=C(Cl)C=C1N

Specifications

70°C to 74°C
C6H7ClN2
4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
NC1=CC=C(Cl)C=C1N
142.59
CHEBI:82301
97%
95-83-0
MFCD00011691
BXIXXXYDDJVHDL-UHFFFAOYSA-N
4-chlorobenzene-1,2-diamine
7263
142.59
4-Chloro-o-phenylenediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350
May cause genetic defects.
May cause cancer.

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335-H350

missing translation for 'einecsNumber' : 202-456-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only