missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals
Solubility
Soluble in water (0.11 mg/ml at 25°C), alcohol, and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases.
Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals
Solubility
Soluble in water (0.11 mg/ml at 25°C), alcohol, and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases.
Specifications
Specifications
Melting Point | 17°C to 20°C |
Density | 1.19 |
Boiling Point | 230°C to 232°C |
Flash Point | 90°C (194°F) |
Refractive Index | 1.555 |
Quantity | 250 g |
UN Number | UN3416 |
Beilstein | 386014 |
Merck Index | 14,2116 |
Solubility Information | Soluble in water (0.11mg/ml at 25°C),alcohol,and ether. |
Show More |
Safety and Handling
Product Identifier
- 4'-Chloroacetophenone
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 4'-Chloroacetophenone
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction