missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals

Product Code. 11430053
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals

Solubility
Soluble in water (0.11 mg/ml at 25°C), alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-91-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000624
InChI Key BUZYGTVTZYSBCU-UHFFFAOYSA-N
Synonym 4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene
PubChem CID 7467
ChEBI CHEBI:27538
IUPAC Name 1-(4-chlorophenyl)ethanone
SMILES CC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 17°C to 20°C
Density 1.19
Boiling Point 230°C to 232°C
Flash Point 90°C (194°F)
Refractive Index 1.555
Quantity 5000 g
UN Number UN3416
Beilstein 386014
Merck Index 14,2116
Solubility Information Soluble in water (0.11mg/ml at 25°C),alcohol,and ether.
Formula Weight 154.6
Percent Purity ≥98%
Chemical Name or Material 4'-Chloroacetophenone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.