missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals

Product Code. 10257263
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-91-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000624
InChI Key BUZYGTVTZYSBCU-UHFFFAOYSA-N
Synonym 4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene
PubChem CID 7467
ChEBI CHEBI:27538
IUPAC Name 1-(4-chlorophenyl)ethanone
SMILES CC(=O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 18.0°C
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 232.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4COCH3
Refractive Index 1.5539 to 1.5559
Quantity 1 kg
Beilstein 07,281
Merck Index 15,2116
Specific Gravity 1.19
Solubility Information Solubility in water: 111 mg/l (25°C).
Formula Weight 154.6
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 4'-Chloroacetophenone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4'-Chloroacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'