missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 68.0°C to 72.0°C |
CAS Min % | 97.5 |
Color | Beige to Brown-Purple |
CAS Max % | 100.0 |
Boiling Point | 232.0°C |
Quantity | 5 g |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Physical Form | Crystalline Solid |
Assay Percent Range | 97.5% min. (GC) |
Show More |
Safety and Handling
Product Identifier
- 4-Chloroaniline
- Danger
- Acute toxicity Category 3
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction