missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11944801
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-88-1
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
InChI Key AVPYQKSLYISFPO-UHFFFAOYSA-N
Synonym p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z
PubChem CID 7726
ChEBI CHEBI:28105
IUPAC Name 4-chlorobenzaldehyde
SMILES C1=CC(=CC=C1C=O)Cl

Specifications

Melting Point >46.0°C
Color White to Yellow
Density 1.1960g/mL
Boiling Point 214.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Linear Formula ClC6H4CHO
Quantity 5 kg
Beilstein 07,235
Specific Gravity 1.196
Solubility Information Solubility in water: 935mg/l (20°C)
Formula Weight 140.57
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 4-Chlorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.