missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 10407190
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-60-2
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00007439
InChI Key ZLYBFBAHAQEEQQ-UHFFFAOYSA-N
Synonym 4-chlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-chloro, p-chlorophenylsulfonyl chloride, p-chlorobenzenesulfonyl chloride, 4-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, p-chloro, p-chlorbenzensulfochlorid, 4-chlorophenylsulfonyl chloride, 4-chlorobenzenesulphonyl chloride, 4-chloro-benzenesulfonyl chloride
PubChem CID 7398
IUPAC Name 4-chlorobenzenesulfonyl chloride
SMILES ClC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 52°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClC6H4SO2Cl
Quantity 5 g
Beilstein 11, 55
Solubility Information Solubility in water: insoluble in water
Formula Weight 211.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzenesulfonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.