missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15475797
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23967-57-9
Molecular Formula C9H5ClO2S
Molecular Weight (g/mol) 212.647
MDL Number MFCD06655155
InChI Key IPAXPERGAMNMIJ-UHFFFAOYSA-N
Synonym 4-chlorobenzo b thiophene-2-carboxylic acid, 4-chloro-benzo b thiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 4-chloro, 4-chloro benzo b thiophene-2-carboxylic acid, 4-chlorobenzothiophene-2-carboxylate, 4-chlorobenzothiophene-2-carboxylic acid, 2-carboxy-4-chlorobenzo b thiophene, 4-chloro-2-benzo b thiophenecarboxylic acid
PubChem CID 3854516
IUPAC Name 4-chloro-1-benzothiophene-2-carboxylic acid
SMILES C1=CC2=C(C=C(S2)C(=O)O)C(=C1)Cl

Specifications

Melting Point ∼233°C (sublimation)
Quantity 1 g
Formula Weight 212.65
Percent Purity 97%
Chemical Name or Material 4-Chlorobenzo[b]thiophene-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'