Learn More
4-Chlorobenzoyl chloride, 98%, Thermo Scientific Chemicals
Description
4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer. 4-Chlorobenzoyl chloride may be used in the following studies Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia, Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene, Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer, Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer. 4-Chlorobenzoyl chloride may be used in the following studies Acylation of benzene using different solid acid catalysts such as dodecatungstophosphoric acid (DTPA), DTPA/K-10 clay, K-10, Amberlite, Amberlyst-15, Indion-130, Filtrol-24 clay and sulfated zirconia, Preparation of 1-(4-chlorobenzoyl)-2, 7-dimethoxynaphthalene, Preparation of 4-chlorobenzoyl CoA, via reaction with CoA in KHCO3 buffer, Preparation of 1-aryloxyacetyl-4-(4-chlorobenzoyl)-semicarbazides.
Solubility
Reacts with water. Reacts with alcohol.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alcohol, bases.

Specifications
Specifications
Melting Point | 12°C to 14°C |
Density | 1.377 |
Boiling Point | 221°C to 222°C |
Flash Point | 105°C (221°F) |
Odor | Acrid |
Refractive Index | 1.578 |
Quantity | 100 g |
UN Number | UN3265 |
Beilstein | 471606 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling


- 4-Chlorobenzoyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 4-Chlorobenzoyl chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.