missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl bromide, 98%, Thermo Scientific Chemicals

Product Code. 10519623
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-95-7
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00040714
InChI Key KQNBRMUBPRGXSL-UHFFFAOYSA-N
Synonym 4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide
PubChem CID 69329
IUPAC Name 1-(bromomethyl)-4-chlorobenzene
SMILES ClC1=CC=C(CBr)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 46°C to 50°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4Cl
Quantity 25 g
Formula Weight 205.48
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Chlorobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'