missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloronicotinic acid, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.
Solubility
Soluble in organic solvents at 20 deg C
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers
4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.
Solubility
Soluble in organic solvents at 20 deg C
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers
Specifications
Specifications
Melting Point | ∽165°C (decomposition) |
Quantity | 1 g |
Beilstein | 9255849 |
Solubility Information | Soluble in organic solvents at 20 deg C |
Formula Weight | 157.56 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chloronicotinic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'