missing translation for 'onlineSavingsMsg'
Learn More

4-Chloronicotinic acid, 96%, Thermo Scientific Chemicals

Product Code. 11324436
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.

Solubility
Soluble in organic solvents at 20 deg C

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10177-29-4
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00128860
InChI Key IMRGVWZLCZERSQ-UHFFFAOYSA-N
Synonym 4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 4-chloro, 3-carboxy-4-chloropyridine, 4-chlornicotinsaure, pubchem12989, acmc-1br3v, 4-chloropyridine-3-carboxyl
PubChem CID 818229
IUPAC Name 4-chloropyridine-3-carboxylic acid
SMILES OC(=O)C1=C(Cl)C=CN=C1

Specifications

Melting Point ∽165°C (decomposition)
Quantity 5 g
Beilstein 9255849
Solubility Information Soluble in organic solvents at 20 deg C
Formula Weight 157.56
Percent Purity 97%
Chemical Name or Material 4-Chloronicotinic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'