missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloronicotinic acid, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Melting Point | 170°C to 171°C |
Assay Percent Range | 95% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 157.56 |
Percent Purity | 96% |
Chemical Name or Material | 4-Chloronicotinic acid |
Safety and Handling
Product Identifier
- 4-Chloronicotinic acid, 96%
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'