missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl methyl sulfone, 98+%, Thermo Scientific™

Product Code. 11488416
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-57-7
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00025067
InChI Key LMCOQDVJBWVNNI-UHFFFAOYSA-N
Synonym 4-chlorophenyl methyl sulfone, 1-chloro-4-methylsulfonyl benzene, p-chlorophenyl methyl sulfone, 4-methylsulfuryl chlorobenzene, benzene, 1-chloro-4-methylsulfonyl, methyl 4-chlorophenyl sulfone, sulfone, p-chlorophenyl methyl, 1-chloro-4-methylsulphonyl benzene, 4-chlorophenylmethyl sulfone, methyl p-chlorophenyl sulfone
PubChem CID 7395
IUPAC Name 1-chloro-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

Melting Point 95°C to 99°C
Quantity 10 g
Beilstein 638281
Formula Weight 190.66
Percent Purity ≥98%
Chemical Name or Material 4-Chlorophenyl methyl sulfone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'