missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10540153
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
AcroSeal™ Glass Bottle in Tin Can
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-77-8, 60-29-7
Molecular Formula C6H4BrClMg
Molecular Weight (g/mol) 215.76
MDL Number MFCD00009926
InChI Key CDEMHJCJMMOFMB-UHFFFAOYSA-M
Synonym 4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent
PubChem CID 101888
IUPAC Name magnesium;chlorobenzene;bromide
SMILES ClC1=CC=C([Mg]Br)C=C1

Specifications

Color Black-Brown to Gray
CAS Min % 22.85, 69.54
CAS Max % 30.46, 77.15
Linear Formula Cl(C6H4)MgBr
Concentration 0.9 to 1.2M
Density 0.8500g/mL
Formula Weight 215.76
Packaging AcroSeal™ Glass Bottle
Quantity 800 mL
Physical Form Solution
Chemical Name or Material 4-Chlorophenylmagnesium bromide, 1M Solution in Diethyl Ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Chlorophenylmagnesium bromide, 1M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'