missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyridine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 10336220 Shop All Thermo Scientific Products
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7379-35-3
Molecular Formula C5H5Cl2N
Molecular Weight (g/mol) 150.00
MDL Number MFCD00012829
InChI Key XGAFCCUNHIMIRV-UHFFFAOYSA-N
Synonym 4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride
PubChem CID 81852
IUPAC Name 4-chloropyridine;hydrochloride
SMILES [H+].[Cl-].ClC1=CC=NC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 210.0°C
Color Beige-Yellow to White
Flash Point 198°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (C5H4N)Cl
Quantity 25 g
Beilstein 20, 231
Solubility Information Solubility in water: soluble
Formula Weight 150.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloropyridine hydrochloride, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloropyridine hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.