missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorotoluene, 98%, Thermo Scientific Chemicals

Product Code. 11381657
Click to view available options
Quantity:
100 g
500 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Chlorotoluene is used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorotoluene is used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

Solubility
Soluble in water (0.04 g/L at 20oc).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-43-4
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000631
InChI Key NPDACUSDTOMAMK-UHFFFAOYSA-N
Synonym 4-chlorotoluene, p-chlorotoluene, benzene, 1-chloro-4-methyl, p-tolyl chloride, toluene, p-chloro, 1-methyl-4-chlorobenzene, 4-chloro-1-methylbenzene, monochlorotoluene, para-chlorotoluene, unii-q8r236h42n
PubChem CID 7810
ChEBI CHEBI:34401
IUPAC Name 1-chloro-4-methylbenzene
SMILES CC1=CC=C(Cl)C=C1

Specifications

Melting Point 7°C to 9°C
Density 1.069
Boiling Point 161°C to 163°C
Quantity 100 g
Odor Aromatic
Refractive Index 1.52
UN Number UN2238
Beilstein 1903635
Merck Index 14,2171
Solubility Information Soluble in water (0.04g/L at 20oc).
Formula Weight 126.59
Percent Purity 98%
Chemical Name or Material 4-Chlorotoluene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.