missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chromanone, 96%, Thermo Scientific Chemicals
77.28 € - 251.25 €
Chemical Identifiers
CAS | 491-37-2 |
---|---|
Molecular Formula | C9H8O2 |
Molecular Weight (g/mol) | 148.16 |
MDL Number | MFCD00006840 |
InChI Key | MSTDXOZUKAQDRL-UHFFFAOYSA-N |
Synonym | 4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone |
PubChem CID | 68110 |
IUPAC Name | 2,3-dihydrochromen-4-one |
SMILES | C1COC2=CC=CC=C2C1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10295860
|
Thermo Scientific Acros
153460100 |
10 g | Glass bottle |
77.28 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10347280
|
Thermo Scientific Acros
153460500 |
50 g | Glass bottle |
251.25 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
491-37-2 | |
148.16 | |
MSTDXOZUKAQDRL-UHFFFAOYSA-N | |
68110 | |
C1COC2=CC=CC=C2C1=O |
C9H8O2 | |
MFCD00006840 | |
4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone | |
2,3-dihydrochromen-4-one |
Spezifikation
491-37-2 | |
100.0 | |
White to Yellow | |
110°C | |
95% min. (GC) | |
1.575 | |
17, II, 335 | |
MSTDXOZUKAQDRL-UHFFFAOYSA-N | |
2,3-dihydrochromen-4-one | |
68110 | |
96% | |
4-Chromanone, 96% |
95.0 | |
34°C to 40°C | |
127°C to 128°C (13.0 mmHg) | |
Authentic | |
C9H8O2 | |
MFCD00006840 | |
4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone | |
C1COC2=CC=CC=C2C1=O | |
148.16 | |
148.16 | |
Low Melting Mass, Crystals or Crystalline Powder |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 207-736-3
missing translation for 'rtecsNumber' : DJ2981225
missing translation for 'tsca' : TSCA
Produktidentifikator
- 4-Chromanone
RUO – Research Use Only