missing translation for 'onlineSavingsMsg'
Learn More

4-CMTB, Tocris Bioscience™

Product Code. 30001592
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

FFA2 agonist; also exhibits positive allosteric modulatory activity

Free fatty acid receptor FFA2 agonist (pEC50 = 6.38) and positive allosteric modulator. Binds at a site distinct from the orthosteric site; modulates the activity of short-chain fatty acids at FFA2 via the FFA2 second extracellular loop (ECL2).

Chemical Identifiers

CAS 300851-67-6
Molecular Formula C14H15ClN2OS
Molecular Weight (g/mol) 294.797
InChI Key AZYDQCGCBQYFSE-UHFFFAOYSA-N
Synonym 4-cmtb, 2-4-chlorophenyl-3-methyl-n-thiazol-2-yl butanamide, 2-4-chlorophenyl-3-methyl-n-1,3-thiazol-2-yl butanamide, enamine_000061, d0i3eq, 4-cmtb hplc, 4-chloro-?-1-methylethyl-n-2-thiazolylbenzeneacetamide, 4-chloro-alpha-1-methylethyl-n-2-thiazolylbenzeneacetamide
PubChem CID 4307629
IUPAC Name 2-(4-chlorophenyl)-3-methyl-N-(1,3-thiazol-2-yl)butanamide
SMILES CC(C)C(C1=CC=C(C=C1)Cl)C(=O)NC2=NC=CS2

Specifications

Quantity 50 mg
Formula Weight 294.8
Percent Purity >98%
Chemical Name or Material 4-CMTB
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'