Learn More
4-Cyano-3-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
67.10 € - 728.00 €
Chemical Identifiers
CAS | 843663-18-3 |
---|---|
Molecular Formula | C7H5BFNO2 |
Molecular Weight (g/mol) | 164.93 |
MDL Number | MFCD03411549 |
InChI Key | DECWLXUOZUMPBF-UHFFFAOYSA-N |
Synonym | 4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug |
PubChem CID | 2783130 |
IUPAC Name | (4-cyano-3-fluorophenyl)boronic acid |
SMILES | OB(O)C1=CC(F)=C(C=C1)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392819
|
Thermo Scientific Alfa Aesar
H27710.03 |
1 g |
67.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302829
|
Thermo Scientific Alfa Aesar
H27710.06 |
5 g | N/A | ||||||
11312829
|
Thermo Scientific Alfa Aesar
H27710.14 |
25 g |
728.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
843663-18-3 | |
164.93 | |
DECWLXUOZUMPBF-UHFFFAOYSA-N | |
2783130 | |
OB(O)C1=CC(F)=C(C=C1)C#N |
C7H5BFNO2 | |
MFCD03411549 | |
4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug | |
(4-cyano-3-fluorophenyl)boronic acid |
Specifications
843663-18-3 | |
C7H5BFNO2 | |
4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug | |
OB(O)C1=CC(F)=C(C=C1)C#N | |
164.93 | |
164.93 | |
4-Cyano-3-fluorobenzeneboronic acid |
>300°C | |
MFCD03411549 | |
DECWLXUOZUMPBF-UHFFFAOYSA-N | |
(4-cyano-3-fluorophenyl)boronic acid | |
2783130 | |
97% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only