Learn More
4-Cyano-4-phenylcyclohexanone, 97%, Thermo Scientific Chemicals
35.78 € - 228.00 €
Chemical Identifiers
CAS | 25115-74-6 |
---|---|
Molecular Formula | C13H13NO |
Molecular Weight (g/mol) | 199.25 |
MDL Number | MFCD00044816 |
InChI Key | GKXOABVSZWCJJK-UHFFFAOYSA-N |
Synonym | 4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone |
PubChem CID | 91282 |
IUPAC Name | 4-oxo-1-phenylcyclohexane-1-carbonitrile |
SMILES | O=C1CCC(CC1)(C#N)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10183230
|
Thermo Scientific Acros
134900050 |
5 g | Glass bottle |
35.78 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10674802
|
Thermo Scientific Acros
134900250 |
25 g | Plastic bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25115-74-6 | |
199.25 | |
GKXOABVSZWCJJK-UHFFFAOYSA-N | |
91282 | |
O=C1CCC(CC1)(C#N)C1=CC=CC=C1 |
C13H13NO | |
MFCD00044816 | |
4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone | |
4-oxo-1-phenylcyclohexane-1-carbonitrile |
Specifications
25115-74-6 | |
100.0 | |
White to Beige | |
97% | |
C6H5(CN)C6H8(=O) | |
4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone | |
GKXOABVSZWCJJK-UHFFFAOYSA-N | |
4-oxo-1-phenylcyclohexane-1-carbonitrile | |
91282 | |
97% | |
4-Cyano-4-phenylcyclohexanone, 97% |
96.0 | |
114°C to 119°C | |
Authentic | |
C13H13NO | |
MFCD00044816 | |
Solubility in water: .. Other solubilities: soluble in benzene | |
O=C1CCC(CC1)(C#N)C1=CC=CC=C1 | |
199.25 | |
199.25 | |
Crystalline Solid |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 246-631-7
- 4-Cyano-4-phenylcyclohexanone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.