missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenyl 4-n-hexylbenzoate, 99%, Thermo Scientific Chemicals

Product Code. 11450647
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50793-85-6
Molecular Formula C20H21NO2
Molecular Weight (g/mol) 307.39
MDL Number MFCD00600506
InChI Key DEUWEGPRKHPNKB-UHFFFAOYSA-N
Synonym benzoic acid, 4-hexyl-, 4-cyanophenyl ester, 4-cyanophenyl 4-n-hexylbenzoate, p-cyanophenyl p-hexylbenzoate, 4-cyanophenyl 4-hexylbenzoate, 4-hexylbenzoic acid 4-cyanophenyl, 4-n-hexylbenzoic acid 4-cyanophenyl ester
PubChem CID 170907
IUPAC Name (4-cyanophenyl) 4-hexylbenzoate
SMILES CCCCCCC1=CC=C(C=C1)C(=O)OC1=CC=C(C=C1)C#N

Specifications

Melting Point 44°C to 47°C
Quantity 5 g
Formula Weight 307.39
Percent Purity 99%
Chemical Name or Material 4-Cyanophenyl 4-n-hexylbenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'