missing translation for 'onlineSavingsMsg'
Learn More
4-(Dimethylamino)pyridine, 99%, prilled, Thermo Scientific Chemicals Product Code.: 15448167

4-(Dimethylamino)pyridine, 99%, prilled, Thermo Scientific Chemicals

Product Code. 15448167
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15448167

Brand: Thermo Scientific Alfa Aesar H51715.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.

Solubility
Soluble in methanol, benzene, ethyl acetate, chloroform, methylene chloride, acetone, and acetic acid. Slightly soluble in ether, diisopropyl ether, cyclohexane, hexane, and water.

Notes
Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-58-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00006418
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
Synonym 4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
PubChem CID 14284
IUPAC Name N,N-dimethylpyridin-4-amine
SMILES CN(C)C1=CC=NC=C1

Specifications

Melting Point 110°C to 113°C
Boiling Point 162°C (50 mmHg)
Flash Point 124°C (255°F)
Quantity 100 g
UN Number UN2928
Beilstein 110354
Solubility Information Soluble in methanol,benzene,ethyl acetate,chloroform,methylene chloride,acetone,and acetic acid. Slightly soluble in ether,diisopropyl ether,cyclohexane,hexane,and water.
Formula Weight 122.17
Percent Purity 99%
Physical Form Prills
Chemical Name or Material 4-(Dimethylamino)pyridine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM31L032 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberH51715.22,H51715.06,H51715.14
Lot NumberQ25D056 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberH51715.22,H51715.06,H51715.14
Lot NumberU25D021 Certificate TypeCertificate of Analysis Date25/05/2022 Catalog NumberH51715.22,H51715.06,H51715.14
Lot NumberG05Y006 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51715.22,H51715.06,H51715.14
Lot NumberP23B015 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51715.22,H51715.06,H51715.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Dimethylamino)pyridine, 99%, prilled, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.