missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminoantipyrine, 97%, Thermo Scientific Chemicals

Product Code. 10274090
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-15-1
Molecular Formula C13H17N3O
Molecular Weight (g/mol) 231.30
MDL Number MFCD00003142
InChI Key RMMXTBMQSGEXHJ-UHFFFAOYSA-N
Synonym aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone
PubChem CID 6009
ChEBI CHEBI:160246
IUPAC Name 4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 105°C to 110°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 231.3
Percent Purity ≥96.0% (Total base)
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Dimethylaminoantipyrine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Dimethylaminoantipyrine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.