missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzaldehyde, ACS, Thermo Scientific Chemicals

Product Code. 11334207
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 73°C to 75°C
Color White to Yellow
Boiling Point 176°C to 177°C (17 mmHg)
Flash Point 164°C (327°F)
Odor Strong
Quantity 25 g
Beilstein 606802
Sensitivity Air and light sensitive
Merck Index 14,3230
Solubility Information Soluble in alcohol,ether,chloroform,acetic acid; Insoluble in water.
Formula Weight 149.19
Grade ACS
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'