missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals

24.12 € - 2075.33 €

Chemical Identifiers

CAS 1122-58-3
MDL Number MFCD00006418
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
Synonym 4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
PubChem CID 14284
IUPAC Name N,N-dimethylpyridin-4-amine
SMILES CN(C)C1=CC=NC=C1
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10504081
SDS View Documents
Thermo Scientific Acros
148270050
5 g Glass bottle
24.12 €
5g
Please to purchase this item. Need a web account? Register with us today!
10091630
SDS View Documents
Thermo Scientific Acros
148270250
25 g Glass bottle
44.62 €
25g
Please to purchase this item. Need a web account? Register with us today!
10133480
SDS View Documents
Thermo Scientific Acros
148271000
100 g Glass Bottle
121.61 €
100g
Please to purchase this item. Need a web account? Register with us today!
10409633
SDS View Documents
Thermo Scientific Acros
148275000
500 g Glass bottle
468.33 €
500g
Please to purchase this item. Need a web account? Register with us today!
11916301
SDS View Documents
Thermo Scientific Acros
148270025
2.5 kg Plastic bottle
2075.33 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.