missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Epioxytetracycline, 'can be used as secondary standard'

Product Code. 10389380
100 mg, Glass bottle
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Epioxytetracycline, CAS # 14206-58-7, is one of the main oxytetracycline metabolites that show antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 4-Epioxytetracycline is a metabolite of the oxytetracycline compound that displays antibacterial activity
  • It is the main degradation product derived from the oxytetracycline metabolization and has been detected in some biological samples
  • Its mechanism of action has been described to be related to the mitochondrial protein synthesis inhibition

Applications

  • In laboratory research it has been described that the treatment with 4-epioxytetracycline decreases the bacterial diversity of the intestinal microbiota and facilitates the proliferation of opportunistic pathogens in salmon
  • The use of 4-epioxytetracycline in animal models (wistar rats) resulted in a significant alteration of the gut microbiota and disorders in blood metabolomics
  • 4-Epioxytetracycline can be applied as a secondary reference standard during stability studies of oxytetracycline
  • 4-epioxytetracycline has been used in research studies to evaluate the degradation pathway and products of tetracyclines
TRUSTED_SUSTAINABILITY

Specifications

CAS 14206-58-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 168.0°C
Color Beige-Green to Brown
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Molecular Formula C22H24N2O9
MDL Number MFCD00062825
Quantity 100 mg
Merck Index 10, 6846
Specific Rotation − 247.00
Synonym 4-epioxytetracyline, unii-73von8sry5, 4-epioxytetracycline, 73von8sry5, 4r,4ar,5s,5ar,6s,12as-4-dimethylamino-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide, hydroxy quatrimycin, oxytetracycline dihydrate specified impurity a ep, oxytetracycline hydrochloride specified impurity a ep, 4r,4ar,5s,5ar,6s,12as-4-dimethylamino-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide, 2-naphthacenecarboxamide, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, 4r,4ar,5s,5ar,6s,12as
Solubility Information Solubility in water: 0.5g/L. Other solubilities: 3.6g/L methanol
InChI Key OWFJMIVZYSDULZ-DVJPNYBFSA-N
SMILES CC1(C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O)O
IUPAC Name (4R,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide
Molecular Weight (g/mol) 460.43
PubChem CID 54678403
Formula Weight 460.43
Percent Purity ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
Physical Form Powder
Chemical Name or Material 4-Epioxytetracycline, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Epioxytetracycline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 4-Epioxytetracycline, 'can be used as secondary standard' >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.