missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,6-difluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 15414767
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Ethoxy-2,6-difluorobenzyl bromide, is used as an intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-51-9
Molecular Formula C9H9BrF2O
Molecular Weight (g/mol) 251.071
MDL Number MFCD09258697
InChI Key FLUSMXIXICUDDW-UHFFFAOYSA-N
Synonym 2-bromomethyl-5-ethoxy-1,3-difluorobenzene, 4-ethoxy-2,6-difluorobenzyl bromide, 2,6-difluoro-4-ethoxybenzyl bromide, 4-ethoxy-2,6-difluoro benzyl bromide
PubChem CID 46737616
IUPAC Name 2-(bromomethyl)-5-ethoxy-1,3-difluorobenzene
SMILES CCOC1=CC(=C(C(=C1)F)CBr)F

Specifications

Melting Point 38°C to 41°C
Quantity 250 mg
UN Number UN3261
Solubility Information Slightly soluble in water.
Formula Weight 251.07
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,6-difluorobenzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.