missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethoxy-3',5'-difluoroacetophenone, 97%, Thermo Scientific™

Product Code. 15484157
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-29-1
Molecular Formula C10H10F2O2
Molecular Weight (g/mol) 200.19
MDL Number MFCD09258688
InChI Key ZKFNHJQYENLQNK-UHFFFAOYSA-N
Synonym 4'-ethoxy-3',5'-difluoroacetophenone, 1-4-ethoxy-3,5-difluorophenyl ethanone, 3',5'-difluoro-4'-ethoxyacetophenone, 1-acetyl-4-ethoxy-3,5-difluorobenzene, 4'-ethoxy-3',5'-difluoroacetophenone, jrd
PubChem CID 46737608
IUPAC Name 1-(4-ethoxy-3,5-difluorophenyl)ethanone
SMILES CCOC1=C(F)C=C(C=C1F)C(C)=O

Specifications

Refractive Index 1.4975
Quantity 1 g
Formula Weight 200.18
Percent Purity 97%
Chemical Name or Material 4'-Ethoxy-3',5'-difluoroacetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.