missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethoxy-3'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 15497387
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851263-13-3
Molecular Formula C11H11F3O2
Molecular Weight (g/mol) 232.202
MDL Number MFCD13196590
InChI Key KWNZFVILRICPQN-UHFFFAOYSA-N
Synonym 4'-ethoxy-3'-trifluoromethyl acetophenone, 1-4-ethoxy-3-trifluoromethyl phenyl ethanone, 1-acetyl-4-ethoxy-3-trifluoromethyl benzene
PubChem CID 57361403
IUPAC Name 1-[4-ethoxy-3-(trifluoromethyl)phenyl]ethanone
SMILES CCOC1=C(C=C(C=C1)C(=O)C)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 232.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4'-Ethoxy-3'-(trifluoromethyl)acetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.