missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals

Product Code. 11493504
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-44-2
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00009094
InChI Key CPJSUEIXXCENMM-UHFFFAOYSA-N
Synonym phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin
PubChem CID 4754
ChEBI CHEBI:8050
IUPAC Name N-(4-ethoxyphenyl)acetamide
SMILES CCOC1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 134°C to 136°C
Quantity 500 g
Beilstein 1869238
Merck Index 14,7204
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 4'-Ethoxyacetanilide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.