missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethoxyacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11468863
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4'-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

Solubility
Soluble in alcohol, water(791.1 mg/L).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1676-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009095
InChI Key YJFNFQHMQJCPRG-UHFFFAOYSA-N
Synonym 4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone
PubChem CID 72872
IUPAC Name 1-(4-ethoxyphenyl)ethanone
SMILES CCOC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 35°C to 38°C
Boiling Point 145°C to 146°C (13 mmHg)
Flash Point 134°C (273°F)
Quantity 5 g
Beilstein 636783
Solubility Information Soluble in alcohol,water(791.1mg/L).
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4'-Ethoxyacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'