missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Ethoxyacetophenone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4'-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.
Solubility
Soluble in alcohol, water(791.1 mg/L).
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
4′-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.
Solubility
Soluble in alcohol, water(791.1 mg/L).
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 35°C to 38°C |
Boiling Point | 145°C to 146°C (13 mmHg) |
Flash Point | 134°C (273°F) |
Quantity | 25 g |
Beilstein | 636783 |
Solubility Information | Soluble in alcohol,water(791.1mg/L). |
Formula Weight | 164.2 |
Percent Purity | 98% |
Chemical Name or Material | 4'-Ethoxyacetophenone |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction