missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethoxyacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11488863
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4'-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

Solubility
Soluble in alcohol, water(791.1 mg/L).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1676-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009095
InChI Key YJFNFQHMQJCPRG-UHFFFAOYSA-N
Synonym 4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone
PubChem CID 72872
IUPAC Name 1-(4-ethoxyphenyl)ethanone
SMILES CCOC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 35°C to 38°C
Boiling Point 145°C to 146°C (13 mmHg)
Flash Point 134°C (273°F)
Quantity 100 g
Beilstein 636783
Solubility Information Soluble in alcohol,water(791.1mg/L).
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4'-Ethoxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.