missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethoxyacetophenone, 99%, Thermo Scientific Chemicals

Product Code. 10444615
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1676-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009095
InChI Key YJFNFQHMQJCPRG-UHFFFAOYSA-N
Synonym 4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone
PubChem CID 72872
IUPAC Name 1-(4-ethoxyphenyl)ethanone
SMILES CCOC1=CC=C(C=C1)C(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 35.0°C to 38.0°C
Boiling Point 268.0°C to 269.0°C (758.0 mmHg)
Flash Point 134°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material 4'-Ethoxyacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Ethoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'