missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethoxycinnamic acid, prediminantly trans, 98+%, Thermo Scientific Chemicals
77.50 € - 266.00 €
Chemical Identifiers
CAS | 2373-79-7 |
---|---|
Molecular Formula | C11H12O3 |
Molecular Weight (g/mol) | 192.214 |
MDL Number | MFCD00016848 |
InChI Key | DZLOUWYGNATKKZ-VMPITWQZSA-N |
Synonym | 4-ethoxycinnamic acid, 3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl prop-2-enoic acid, e-3-4-ethoxyphenyl acrylic acid, e-3-4-ethoxyphenyl prop-2-enoic acid, 4-ethoxyphenylacrylic acid, p-ethoxycinnamic acid, ethoxyphenylacrylicacid, pubchem8239 |
PubChem CID | 704218 |
IUPAC Name | (E)-3-(4-ethoxyphenyl)prop-2-enoic acid |
SMILES | CCOC1=CC=C(C=C1)C=CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392456
|
Thermo Scientific Alfa Aesar
L06324.06 |
5 g |
77.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302466
|
Thermo Scientific Alfa Aesar
L06324.14 |
25 g |
266.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2373-79-7 | |
192.214 | |
DZLOUWYGNATKKZ-VMPITWQZSA-N | |
704218 | |
CCOC1=CC=C(C=C1)C=CC(=O)O |
C11H12O3 | |
MFCD00016848 | |
4-ethoxycinnamic acid, 3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl prop-2-enoic acid, e-3-4-ethoxyphenyl acrylic acid, e-3-4-ethoxyphenyl prop-2-enoic acid, 4-ethoxyphenylacrylic acid, p-ethoxycinnamic acid, ethoxyphenylacrylicacid, pubchem8239 | |
(E)-3-(4-ethoxyphenyl)prop-2-enoic acid |
Specifications
2373-79-7 | |
C11H12O3 | |
3198068 | |
DZLOUWYGNATKKZ-VMPITWQZSA-N | |
(E)-3-(4-ethoxyphenyl)prop-2-enoic acid | |
704218 | |
≥98% |
195°C to 199°C | |
MFCD00016848 | |
4-ethoxycinnamic acid, 3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl prop-2-enoic acid, e-3-4-ethoxyphenyl acrylic acid, e-3-4-ethoxyphenyl prop-2-enoic acid, 4-ethoxyphenylacrylic acid, p-ethoxycinnamic acid, ethoxyphenylacrylicacid, pubchem8239 | |
CCOC1=CC=C(C=C1)C=CC(=O)O | |
192.214 | |
192.22 | |
4-Ethoxycinnamic acid, predominantly trans |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only