missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11453148
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3460-49-9
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00041096
InChI Key REEKOMRZYJXXNR-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate
PubChem CID 77006
IUPAC Name 1-ethoxy-4-isothiocyanatobenzene
SMILES CCOC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 57°C to 61°C
Flash Point >110°C (230°F)
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 637526
Sensitivity Moisture sensitive
Formula Weight 179.24
Percent Purity 98%
Chemical Name or Material 4-Ethoxyphenyl isothiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'