missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11463148
Click to view available options
Quantity:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 3460-49-9
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00041096
InChI Key REEKOMRZYJXXNR-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate
PubChem CID 77006
IUPAC Name 1-ethoxy-4-isothiocyanatobenzene
SMILES CCOC1=CC=C(C=C1)N=C=S

Spécification

Melting Point 57°C to 61°C
Flash Point >110°C (230°F)
Odor Irritating
Quantity 25 g
UN Number UN2923
Beilstein 637526
Sensitivity Moisture sensitive
Formula Weight 179.24
Percent Purity 98%
Chemical Name or Material 4-Ethoxyphenyl isothiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'