Learn More
4-Ethylbenzoyl chloride, 97%, Thermo Scientific Chemicals
45.12 € - 154.77 €
Chemical Identifiers
CAS | 16331-45-6 |
---|---|
Molecular Formula | C9H9ClO |
Molecular Weight (g/mol) | 168.62 |
MDL Number | MFCD00000697 |
InChI Key | AVTLLLZVYYPGFX-UHFFFAOYSA-N |
Synonym | benzoyl chloride, 4-ethyl, p-ethylbenzoyl chloride, 4-ethylbenzoylchloride, 4-ethylbenzene-1-carbonyl chloride, 4-ethyl-benzoyl chloride, acmc-20amiy, pubchem9725, 4-ethyl benzoyl choride, 4-ethyl benzoyl chloride, benzoyl chloride,4-ethyl |
PubChem CID | 85368 |
IUPAC Name | 4-ethylbenzoyl chloride |
SMILES | CCC1=CC=C(C=C1)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10093740
|
Thermo Scientific Acros
305730050 |
5 g | Glass bottle |
45.12 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10761735
|
Thermo Scientific
305730250 |
25 g | Glass bottle |
154.77 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16331-45-6 | |
168.62 | |
AVTLLLZVYYPGFX-UHFFFAOYSA-N | |
85368 | |
CCC1=CC=C(C=C1)C(=O)Cl |
C9H9ClO | |
MFCD00000697 | |
benzoyl chloride, 4-ethyl, p-ethylbenzoyl chloride, 4-ethylbenzoylchloride, 4-ethylbenzene-1-carbonyl chloride, 4-ethyl-benzoyl chloride, acmc-20amiy, pubchem9725, 4-ethyl benzoyl choride, 4-ethyl benzoyl chloride, benzoyl chloride,4-ethyl | |
4-ethylbenzoyl chloride |
Specifications
16331-45-6 | |
100.0 | |
1.1400g/mL | |
>110°C | |
96% min. (GC) | |
C2H5C6H4COCl | |
MFCD00000697 | |
benzoyl chloride, 4-ethyl, p-ethylbenzoyl chloride, 4-ethylbenzoylchloride, 4-ethylbenzene-1-carbonyl chloride, 4-ethyl-benzoyl chloride, acmc-20amiy, pubchem9725, 4-ethyl benzoyl choride, 4-ethyl benzoyl chloride, benzoyl chloride,4-ethyl | |
CCC1=CC=C(C=C1)C(=O)Cl | |
168.62 | |
168.62 | |
Liquid |
96.0 | |
Yellow | |
235°C to 236°C | |
Authentic | |
C9H9ClO | |
1.5465 to 1.5485 | |
1.14 | |
AVTLLLZVYYPGFX-UHFFFAOYSA-N | |
4-ethylbenzoyl chloride | |
85368 | |
97% | |
4-Ethylbenzoyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 240-404-6
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.