Learn More
4-Ethylbenzoyl chloride, 98+%, Thermo Scientific Chemicals
49.50 € - 300.00 €
Chemical Identifiers
CAS | 16331-45-6 |
---|---|
Molecular Formula | C9H9ClO |
Molecular Weight (g/mol) | 168.62 |
MDL Number | MFCD00000697 |
InChI Key | AVTLLLZVYYPGFX-UHFFFAOYSA-N |
PubChem CID | 85368 |
IUPAC Name | 4-ethylbenzoyl chloride |
SMILES | CCC1=CC=C(C=C1)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324739
|
Thermo Scientific Alfa Aesar
A11698.06 |
5 g |
49.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419213
|
Thermo Scientific Alfa Aesar
A11698.09 |
10 g |
80.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429213
|
Thermo Scientific Alfa Aesar
A11698.18 |
50 g |
300.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16331-45-6 | |
168.62 | |
AVTLLLZVYYPGFX-UHFFFAOYSA-N | |
4-ethylbenzoyl chloride |
C9H9ClO | |
MFCD00000697 | |
85368 | |
CCC1=CC=C(C=C1)C(=O)Cl |
Specifications
16331-45-6 | |
>110°C (230°F) | |
C9H9ClO | |
UN3265 | |
benzoyl chloride, 4-ethyl, p-ethylbenzoyl chloride, 4-ethylbenzoylchloride, 4-ethylbenzene-1-carbonyl chloride, 4-ethyl-benzoyl chloride, acmc-20amiy, pubchem9725, 4-ethyl benzoyl choride, 4-ethyl benzoyl chloride, benzoyl chloride,4-ethyl | |
CCC1=CC=C(C=C1)C(=O)Cl | |
168.62 | |
168.62 | |
4-Ethylbenzoyl chloride |
235°C to 236°C | |
Acidic | |
MFCD00000697 | |
2206041 | |
AVTLLLZVYYPGFX-UHFFFAOYSA-N | |
4-ethylbenzoyl chloride | |
85368 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
RUO – Research Use Only