missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenol, 97%

Product Code. 10306560 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5g
5kg
This item is not returnable. View return policy

Product Code. 10306560

Brand: Thermo Scientific undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-07-9 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-07-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
InChI Key HXDOZKJGKXYMEW-UHFFFAOYSA-N
Synonym p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol
PubChem CID 31242
ChEBI CHEBI:49584
IUPAC Name 4-ethylphenol
SMILES CCC1=CC=C(C=C1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 41.0°C to 46.0°C
Color Brown to White
Boiling Point 218.0°C to 219.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula C2H5C6H4OH
Quantity 5 g
Beilstein 06,472
Solubility Information Solubility in water: 4900mg/l (25°C). Other solubilities: soluble in alcohol and ether
Formula Weight 122.17
Percent Purity 97%
Physical Form Crystalline Mass or Crystals
Chemical Name or Material 4-Ethylphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Ethylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.