missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

54.07 € - 200.00 €

Chemical Identifiers

CAS 208173-21-1
Molecular Formula C9H6F4O
Molecular Weight (g/mol) 206.14
MDL Number MFCD00061251
InChI Key WBCCAINPZLAKRN-UHFFFAOYSA-N
Synonym 4'-fluoro-2'-trifluoromethyl acetophenone, 1-4-fluoro-2-trifluoromethyl phenyl ethanone, 4-fluoro-2-trifluoromethyl acetophenone, 1-4-fluoro-2-trifluoromethyl phenyl ethan-1-one, ethanone, 1-4-fluoro-2-trifluoromethyl phenyl, 1-acetyl-4-fluoro-2-trifluoromethyl benzene, pubchem4243, acmc-1cmiy, 2-acetyl-5-fluorobenzotrifluoride
PubChem CID 2737535
IUPAC Name 1-[4-fluoro-2-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(F)C=C1C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488937
View Documents
Thermo Scientific Alfa Aesar
B20558.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11498937
View Documents
Thermo Scientific Alfa Aesar
B20558.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

208173-21-1
206.14
WBCCAINPZLAKRN-UHFFFAOYSA-N
2737535
CC(=O)C1=CC=C(F)C=C1C(F)(F)F
C9H6F4O
MFCD00061251
4'-fluoro-2'-trifluoromethyl acetophenone, 1-4-fluoro-2-trifluoromethyl phenyl ethanone, 4-fluoro-2-trifluoromethyl acetophenone, 1-4-fluoro-2-trifluoromethyl phenyl ethan-1-one, ethanone, 1-4-fluoro-2-trifluoromethyl phenyl, 1-acetyl-4-fluoro-2-trifluoromethyl benzene, pubchem4243, acmc-1cmiy, 2-acetyl-5-fluorobenzotrifluoride
1-[4-fluoro-2-(trifluoromethyl)phenyl]ethanone

Specifications

208173-21-1
1.449
7637196
WBCCAINPZLAKRN-UHFFFAOYSA-N
1-[4-fluoro-2-(trifluoromethyl)phenyl]ethanone
2737535
97%
C9H6F4O
MFCD00061251
4'-fluoro-2'-trifluoromethyl acetophenone, 1-4-fluoro-2-trifluoromethyl phenyl ethanone, 4-fluoro-2-trifluoromethyl acetophenone, 1-4-fluoro-2-trifluoromethyl phenyl ethan-1-one, ethanone, 1-4-fluoro-2-trifluoromethyl phenyl, 1-acetyl-4-fluoro-2-trifluoromethyl benzene, pubchem4243, acmc-1cmiy, 2-acetyl-5-fluorobenzotrifluoride
CC(=O)C1=CC=C(F)C=C1C(F)(F)F
206.14
206.14
4'-Fluoro-2'-(trifluoromethyl)acetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.