missing translation for 'onlineSavingsMsg'
Learn More
4-Fluoro-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 11385485

4-Fluoro-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11385485
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11385485

Brand: Thermo Scientific Alfa Aesar L19671.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128495-46-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143320
InChI Key NALVGTOMKSKFFV-UHFFFAOYSA-N
Synonym 3-methoxy-4-fluorobenzaldehyde, 4-fluoro-m-anisaldehyde, 4-fluoro-3-methoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-methoxy, pubchem1449, acmc-209bf6, ksc490m2d, 3-methoxy-4-fluoro benzaldehyde, 3-methoxy-4-fluoro-benzaldehyde, nalvgtomkskffv-uhfffaoysaShow MoreShow Less
PubChem CID 2737358
IUPAC Name 4-fluoro-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1F

Specifications

Melting Point 61°C to 63°C
Quantity 5 g
Beilstein 5178063
Sensitivity Air Sensitive
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-methoxybenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10205100 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog NumberL19671.03,L19671.06
Lot Number10111780 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19671.03,L19671.06
Lot Number10174166 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19671.03,L19671.06
Lot Number10174713 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19671.03,L19671.06
Lot Number10202192 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19671.03,L19671.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluoro-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.